Skip to Content
Merck
5-Methyl-5-phenylhydantoin 99%

5-Methyl-5-phenylhydantoin

5-Methyl-5-phenylhydantoin 99%
Empirical Formula (Hill Notation):
C10H10N2O2
CAS Number:
Molecular Weight:
190.20

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Separation Technique
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing