Skip to Content
Merck
4-Methylumbelliferyl 4-guanidinobenzoate hydrochloride hydrate ≥85% (HPLC)

4-Methylumbelliferyl 4-guanidinobenzoate hydrochloride hydrate

4-Methylumbelliferyl 4-guanidinobenzoate hydrochloride hydrate ≥85% (HPLC)
Synonym(s):
4-Guanidinobenzoic acid 4-methylumbelliferyl ester hydrochloride, MUGB
Empirical Formula (Hill Notation):
C18H15N3O4 · HCl · xH2O
CAS Number:
Molecular Weight:
373.79 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing