Skip to Content
Merck
Crystal violet (C.I. 42555) for microscopy Certistain®

Crystal violet (C.I. 42555)

Crystal violet (C.I. 42555) for microscopy Certistain®
Synonym(s):
Crystal violet (C.I. 42555)
Empirical Formula (Hill Notation):
C25H30ClN3
CAS Number:
Molecular Weight:
407.98

Products

Brand
Formula Weight
Physical Form
Agency
Chemical Composition
Quality Segment
Application
Showing 1-3 of 3