
3-sn-Phosphatidylethanolamine solution

Synonym(s):
L-α-Cephalin
CAS Number:
EC Number:
Products
Product Category
Brand
Biological Source
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing