Skip to Content
Merck
3-Amino-2-naphthoic acid technical grade, 80%

3-Amino-2-naphthoic acid

3-Amino-2-naphthoic acid technical grade, 80%
Linear Formula:
H2NC10H6CO2H
CAS Number:
Molecular Weight:
187.19
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing