Skip to Content
Merck
3,3-Bis(aminoethyl)-1-hydroxy-2-oxo-1-triazene ≥95%

3,3-Bis(aminoethyl)-1-hydroxy-2-oxo-1-triazene

3,3-Bis(aminoethyl)-1-hydroxy-2-oxo-1-triazene ≥95%
Synonym(s):
2,2′-(Hydroxynitrosohydrazino)bis-ethanamine, DETA NONOate, NOC-18
Empirical Formula (Hill Notation):
C4H13N5O2
CAS Number:
Molecular Weight:
163.18

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing