Skip to Content
Merck
2-Methylimidazole (Ondansetron Impurity F) Pharmaceutical Secondary Standard; Certified Reference Material

2-Methylimidazole (Ondansetron Impurity F)

2-Methylimidazole (Ondansetron Impurity F) Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
2-Methylimidazole, 2-Methylglyoxaline
Empirical Formula (Hill Notation):
C4H6N2
CAS Number:
Molecular Weight:
82.10
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique