
2-Linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine

Synonym(s):
1-Hexadecanoyl-2-(cis-9,12-octadecadienoyl)-sn-glycero-3-phosphocholine, 3-sn-Phosphatidylcholine, 2-linoleoyl-1-palmitoyl
Empirical Formula (Hill Notation):
C42H80NO8P
CAS Number:
Molecular Weight:
758.06
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing