Skip to Content
Merck
2-Amino-4-methyl-3-nitropyridine 98%

2-Amino-4-methyl-3-nitropyridine

2-Amino-4-methyl-3-nitropyridine 98%
Synonym(s):
2-Amino-3-nitro-4-picoline
Empirical Formula (Hill Notation):
C6H7N3O2
CAS Number:
Molecular Weight:
153.14

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing