Skip to Content
Merck
2,4,5-Trichlorophenoxyacetic acid suitable for plant cell culture, BioReagent, ≥95%, crystalline

2,4,5-Trichlorophenoxyacetic acid

2,4,5-Trichlorophenoxyacetic acid suitable for plant cell culture, BioReagent, ≥95%, crystalline
Synonym(s):
2,4,5-T
Linear Formula:
Cl3C6H2OCH2CO2H
CAS Number:
Molecular Weight:
255.48
EC Number:

Products

Brand
Biological Source
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Technique
Showing 1-1 of 1