Octane-1-sulfonic acid sodium salt
Synonym(s):
Octane-1-sulfonic acid sodium salt, Sodium 1-octanesulfonate
Empirical Formula (Hill Notation):
C8H17NaO3S
CAS Number:
Molecular Weight:
216.27
Products
Brand
Formula Weight
Physical Form
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing