Skip to Content
Merck
16α,17α-Epoxypregnenolone acetate ~95%

16α,17α-Epoxypregnenolone acetate

16α,17α-Epoxypregnenolone acetate ~95%
Synonym(s):
16α,17-Epoxy-3β-hydroxy-5-pregnen-20-one acetate, 3β-Acetoxy-16α,17α-epoxy-5-pregnen-20-one
Empirical Formula (Hill Notation):
C23H32O4
CAS Number:
Molecular Weight:
372.50
EC Number:

Products

Product Category
Brand
Formula Weight
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing