Skip to Content
Merck
Polymyxin B sulfate meets USP testing specifications

Polymyxin B sulfate salt

Polymyxin B sulfate meets USP testing specifications
Synonym(s):
Polymyxin B sulfate salt, Polymyxin B Sulfate
Empirical Formula (Hill Notation):
C55H96N16O13 · 2H2SO4
CAS Number:
Molecular Weight:
1385.61

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Physical Form
Product Line
Activity Spectrum
Agency
Quality Segment
Application
Mode of Action
Technique