MilliporeSigma
HomeChemical Analysis for Food and Beverage TestingTrans Fats in Cookies using SP-2560 and SLB-IL111
Trans Fats in Cookies using SP-2560 and SLB-IL111
Katherine K. Stenerson
Reporter US Volume 30.2
Trans Fat Labeling
Trans fats (trans unsaturated fatty acids) are fatty acids that contain at least one double bond in the trans orientation. This orientation causes carbon atoms to align in a straight configuration, similar to saturated fatty acids. Most naturally occurring unsaturated fatty acids have double bonds in the cis orientation, which is a bent configuration. Trans fats do not occur naturally in plant-based oils and fats. However, they can be produced commercially through a hydrogenation process, which is used to prevent foods from spoiling, and to produce specific structure effects1. Examples of the structures of saturated, trans unsaturated, and cis unsaturated fatty acids are shown in Figure 1.
Fatty Acid Structures
Figure 1. Fatty Acid Structures
Concerns have been raised for several decades that the consumption of trans fats may have contributed to an epidemic of coronary heart disease1. In response, many government agencies instituted requirements that trans fat content must be listed on the nutritional labels of foods and some dietary supplements. Some regulations state if a food contains less than 0.5 g of trans fat per serving, then “0 g” is allowed to be listed on the label2. If partially hydrogenated fats/oils are used in making the product, small amounts of trans fat will still be present (below 0.5 g even though 0 g is stated on the label).
Extraction and Analysis Methodologies
There are many references available for the preparation of samples for trans fat analysis. In 2007, the American Oil Chemists Society (AOCS®) published method Ce 1k-09 for the preparation of a variety of matrices for the analysis of lipids, including trans fats. This method describes the use of an acid digestion and alkali hydrolysis to release the fats and oils from the sample matrix, followed by methylation using boron trifluoridemethanol (BF3-methanol)3. The fats are then analyzed by GC as their corresponding fatty acid methyl esters (FAMEs). The GC analysis method is chosen based on the sample matrix. In the case of vegetable oils and fats, AOCS Method Ce 1h-05 can be used. This method outlines the use of a 100 m highly polar cyanopropyl silicone capillary column for the single analysis of trans isomers, saturated, cis/trans monounsaturated, and cis/trans polyunsaturated fatty acids4. The length and high polarity of the column are necessary to facilitate resolution of the cis and trans monounsaturates in the oleic (C18:1) region.
Silver-Ion SPE (Ag-Ion SPE) Fractionation
Silver-ion (argentation) chromatography was originally pioneered for lipid analysis by Morris, who demonstrated the first practical applications by separating lipids based on degrees of unsaturation using TLC5. In Ag-Ion SPE, silver ions are anchored onto strong cation exchange (SCX) phase functional groups through electrostatic interaction. As the FAME sample passes through, the SCX-silver counter-ions form specific polar complexes with double bonds of unsaturated FAMEs. The strength of the interaction increases with the number of double bonds, so saturated FAMEs are poorly retained. For unsaturated FAMEs, cis are retained more strongly than trans due to their greater steric accessibility to the SCX-silver ion phase. These differences in interaction strength are then used to separate FAMEs by degree of unsaturation, and also by cis/trans configuration.
In this application, a sample of commercially purchased cookies was extracted and methylated using the method described in AOCS Ce 1k-09. The extract was then fractionated using Discovery® Ag-Ion SPE, and the fractions were analyzed on two 100 m capillary columns, the highly polar SP™-2560 and the extremely polar SLB®-IL111.
Extraction of Cookie Sample
A sample of commercially purchased cookies was ground, and a 1 g sample was subjected to acid digestion and alkali hydrolysis followed by methylation, as described in AOCS Official Method Ce 1k-093. A summary of the extraction and derivatization process is described in Table 1.
An aliquot of the resulting extract was then fractionated using Discovery Ag-ion SPE. The volume of the extract fractionated was calculated to deliver 1 mg of fat to the SPE cartridge. This calculation was made based on nutritional content reported on the product label, and the sample size and solvent volumes used for extraction. After fractionation, samples were dried and reconstituted in 1 mL of hexane for GC-FID analysis. Table 2 contains a summary of the fractionation procedure.
Table 1. Summary of Extraction and Derivatization
  1. Homogenize sample and weigh 1 g into a 50 mL round bottom flask. Add several boiling chips.
  2. Add 5 mL of 1.3 M methanolic hydrochloride (HCl). Attach condenser and reflux for 15 min after mixture has started to boil.
  3. Turn off heat, remove condenser and add 5 mL of 2.3 M methanolic sodium hydroxide (NaOH) to flask. Reattach condenser and reflux again for 15 min.
  4. Turn off heat, remove condenser, and add 14 mL of 10% BF3- methanol. Reattach condenser and reflux for 5 min.
  5. Turn off heat, remove condenser, and add 5 mL of hexane. Remove flask from mantle and let it cool to room temperature.
  6. Add saturated sodium chloride (salt) solution (aqueous) to the flask until organic layer (top) is in the neck.
  7. Stopper the flask and shake. Remove top later, pass through sodium sulfate bed and save for fractionation using Ag-Ion SPE.
Table 2. Summary of Ag-Ion SPE Cleanup
  1. Condition Ag-Ion SPE tube with 4 mL of acetone and allow solvent to drip completely through the tube; discard eluant.
  2. Equilibrate Ag-Ion SPE tube by passing 4 mL of hexane completely through the tube; discard eluant.
  3. Load 1 mL of sample extract onto the tube. Any eluant coming through the tube may be discarded.
  4. Elute fraction 1 with 6 mL of hexane:acetone (96:4) and collect eluant. Be sure all solvent has been drawn through the tube. This fraction should contain any saturates, trans monoenes, and cis/cis and trans/trans conjugated linoleic acids (CLAs).
  5. Elute fraction 2 with 4 mL of hexane: acetone (90:10) and collect eluant. Be sure all solvent has been drawn through the tube. This fraction should contain any cis monenes, trans/trans dienes, and cis/trans and trans/cis CLAs.
  6. Elute fraction 3 with 4 mL of 100% acetone. This fraction should contain any cis/cis dienes, other dienes, and most trienes.
  7. Dry each fraction down at room temperature using nitrogen, and reconstitute in 1 mL of hexane for GC analysis.
Note: Gravity should be sufficient for each step, with vacuum applied briefly only at the end of steps 4, 5, and 6, to help draw the elution solvent completely out of the SPE tube.
Analysis on 100 m SP-2560 and SLB-IL111 Columns
The established AOCS method for analysis of fats in vegetablebased oils and fats requires GC analysis isothermally at 180 °C. This temperature has been determined to provide optimal resolution on a cyanopropyl silicone column (such as the SP-2560) of the C18 FAME isomers usually found in partially hydrogenated vegetable oils6. The SLB-IL111 is made from an ionic liquid stationary phase, and has demonstrated selectivity characteristics indicating it to be more polar than the SP-2560. Studies of cis/trans FAMEs on this column determined 168 °C as the optimal operating temperature for this application6.
Chromatograms of unfractionated cookie extract, and each of the three fractions collected using Ag-Ion SPE, are presented in Figure 2 (SP-2560) and Figure 3 (SLB-IL111). Peaks were identified using retention time and direct comparison to standards. As shown, Ag-ion SPE fractionation was able to separate the saturates and C18 trans monoenes into fraction 1, the cis monoenes into fraction 2, and the dienes into fraction 3.
The nutritional label for the cookies included partially hydrogenated cottonseed and soybean oils as ingredients. Based on this information, the fatty acid profile obtained for this sample was as expected; with cottonseed oil accounting for the small amount of C14:0 FAME, and the soybean oil as the source of the small amount of C22:0 FAME detected. Both oils contributed to the higher levels of the C16:0, C18:0 and C18:1 FAMEs. Note the presence of the C18:1 trans isomers in fraction 1. These are the result of the partial hydrogenation of these oils.
Differences were observed in the elution patterns of the C18:1 cis and trans isomers between the two columns. For example, in Ag- Ion SPE fraction 1, C18:1Δ11t was better resolved from C18:1Δ12t on the SP-2560. In the unfractionated sample, C20:0 elutes after C18:2n6c on the SP-2560 and before on the SLB-IL111. This is due to the stronger dipole-induced dipole interaction exhibited by the SLB-IL111. At the same time, the weaker dispersive interaction demonstrated by the higher polarity SLB-IL111 resulted in faster overall elution of the FAMEs compared to the SP-2560.
SP-2560 Chromatograms
Figure 2. SP-2560 Chromatograms
GC Conditions
sample/matrix: 1 g of commercially purchased cookies was ground and subjected to acid digestion and alkali hydrolysis, followed by methylation as described in AOCS Official Method Ce 1k-09
SPE tube: Discovery Ag-Ion SPE tubes, 750 mg/6 mL (Product No. 54225-U)
conditioning: 4 mL of acetone; allow solvent to gravity drip completely through tube; discard eluant; 4 mL of hexane; allow solvent to gravity drip completely through tube; discard eluant sample addition: 1 mL of extract; discard any eluant that drips through tube elution: (Fraction 1) 6 mL of hexane:acetone (96:4); collect eluant in a fresh container with slight vacuum; (Fraction 2) 4 mL of hexane: acetone (90:10); collect eluant in a fresh container with slight vacuum; (Fraction 3) 4 mL of 100% acetone; collect eluant in a fresh container with slight vacuum eluate: post-treatment: evaporate each fraction at room temperature using nitrogen; reconstitute each fraction to 1 mL of hexane.
column: SP-2560, 100 m x 0.25 mm I.D., 0.20 µm (Product No. 24056) oven: 180 °C; inj. temp.: 250 °C; detector: FID, 250 °C; carrier gas: hydrogen, 1 mL/min; injection: 1 µL, 10:1; split liner: 4 mm I.D., split type, single taper wool packed FocusLiner™ design
SLB-IL111 Chromatograms
Figure 3. SLB-IL111 Chromatograms
GC Conditions
column: SLB-IL111, 100 m x 0.25 mm I.D., 0.20 µM (Product No. 29647-U) oven: 168 °C
Other conditions are the same as Figure 2.
Conclusion
In this application, we have demonstrated the extraction of lipids as fatty acids, and subsequent detection of trans fats from a cookie sample made using partially hydrogenated vegetable oils. Ag-Ion SPE was used to fractionate the extracted sample into saturated FAMEs, and trans monoenes, cis monoenes, and dienes. Subsequent analysis of the monoene fractions on a highly polar SP-2560 column and an extremely polar SLB-IL111 column provided resolution of many of the C18:1 cis/trans geometricpositional isomers, with different elution patterns observed. The columns should be considered complimentary in the analysis of these isomers, with application- specific resolutions considered when choosing which one to use.
Legal Information
Discovery and SLB are registered trademarks of Sigma-Aldrich Co. LLC
SP is a trademark of Sigma-Aldrich Co. LLC
AOCS is a registered trademark of The American Oil Chemists' Society
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
Materials
Loading
References
1.
Ascherio A, Stampfer M, Willet W. 1999. Trans Fatty Acid and Coronary Heart Disease. [Internet]. Available from: http://www.drtimdelivers.com/EEasy122605/Harvardtransfats/transfats
2.
Electronic Code of Federal Regulations; 21 CFR 101.9 (c)(2)(ii). [Internet]. Available from: ecfr. gpoaccess.gov
3.
2011-2012 Method Additions and Revisions. AOCS Official Method Ce 1k-09. Direct Methylation of Lipids in Foods for the Determination of Total Fat, Saturated, cis-Monounsaturated, cis-Polyunsaturated and trans Fatty Acids by Gas Chromatography AOCS Official Methods and Recommended Practices. [Internet]. Available from: https://www.aafco.org/Portals/0/SiteContent/Regulatory/Committees/Lab-Methods-and-Services/Methods/fat_analysis_aocs.pdf
4.
AOCS Official Method Ce 1h-05. Determination of cis-, trans-, Saturated, Monounsaturated, and Polyunsaturated Fatty Acids in Vegetable or Nonruminant Animal Oils and Fats by Capillary GLC. AOCS Official Methods and Recommended Practices of the AOCS. [Internet]. Available from: https://www.aocs.org/attain-lab-services/methods/methods/method-detail?productId=111777
5.
Morris L. 1966. Separations of Lipids by Silver Ion Chromatography J. Lipid Research 7 717-732. [Internet].
6.
Delmonte P, Fardin Kia A, Kramer JK, Mossoba MM, Sidisky L, Rader JI. 2011. Separation characteristics of fatty acid methyl esters using SLB-IL111, a new ionic liquid coated capillary gas chromatographic column. Journal of Chromatography A. 1218(3):545-554. http://dx.doi.org/10.1016/j.chroma.2010.11.072
7.
Buchanan M, Stenerson K, Sidisky L. SLB-IL111 for Fatty Acid Methyl Ester (FAME) Applications Supelco Technical Report (T411139 ODZ). [Internet]. Available from: https://www.sigmaaldrich.com/content/dam/sigma-aldrich/docs/Supelco/General_Information/1/t411139.pdf
8.
Discovery Ag-Ion SPE for FAME Fractionation and Cis/Trans Separation Supelco Technical Report (T406062 IRV). Available from: https://www.sigmaaldrich.com/content/dam/sigma-aldrich/docs/Supelco/Bulletin/t406062.pdf