  • Search Results
  • Neutralization of interleukin-18 ameliorates ischemia/reperfusion-induced myocardial injury.
Neutralization of interleukin-18 ameliorates ischemia/reperfusion-induced myocardial injury.
The Journal of biological chemistry (2009-01-24)
Kaliyamurthi Venkatachalam, Sumanth D Prabhu, Venkatapuram Seenu Reddy, William H Boylston, Anthony J Valente, Bysani Chandrasekar
ABSTRACT
Ischemia/reperfusion (I/R) injury is characterized by the induction of oxidative stress and proinflammatory cytokine expression. Recently demonstrating that oxidative stress and TNF-alpha each stimulate interleukin (IL)-18 expression in cardiomyocytes, we hypothesized that I/R also induces IL-18 expression and thus exacerbates inflammation and tissue damage. Neutralization of IL-18 signaling should therefore diminish tissue injury following I/R. I/R studies were performed using a chronically instrumented closed chest mouse model. Male C57BL/6 mice underwent 30 min of ischemia by LAD coronary artery ligation followed by various periods of reperfusion. Sham-operated or ischemia-only mice served as controls. A subset of animals was treated with IL-18-neutralizing antibodies 1 h prior to LAD ligation. Ischemic LV tissue was used for analysis. Our results demonstrate that, compared with sham operation and ischemia alone, I/R significantly increased (i) oxidative stress (increased MDA/4-HNE levels), (ii) neutrophil infiltration (increased MPO activity), (iii) NF-kappaB DNA binding activity (p50, p65), and (iv) increased expression of IL-18Rbeta, but not IL-18Ralpha or IL-18BP transcripts. Administration of IL-18-neutralizing antibodies significantly reduced I/R injury measured by reduced infarct size (versus control IgG). In isolated adult mouse cardiomyocytes, simulated ischemia/reperfusion enhanced oxidative stress and biologically active IL-18 expression via IKK-dependent NF-kappaB activation. These results indicate that IL-18 plays a critical role in I/R injury and thus represents a promising therapeutic target.
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)