MilliporeSigma
1,25(OH)
Experimental cell research (2017-12-03)
Hua Yu, Min Xu, Yizhi Dong, Jingjing Liu, Yaozhe Li, Wei Mao, Jindan Wang, Liangxing Wang
ABSTRACT
Pulmonary arterial hypertension (PAH) is a devastating disease characterized by high pulmonary artery pressure. It is reported that microRNA-204 (miR-204) plays an important role in the development of PAH. Calcitriol [1,25-dihydroxyvitamin D3, 1,25(OH)