  • Search Results
  • Sensitization and desensitization to allyl isothiocyanate (mustard oil) in the nasal cavity.
Sensitization and desensitization to allyl isothiocyanate (mustard oil) in the nasal cavity.
Chemical Senses (2002)
Brand G & Jacquot L.