  • Search Results
  • Volatile Phoducts of Apples III. Identification of Aldehydes and Ketones.
Volatile Phoducts of Apples III. Identification of Aldehydes and Ketones.
Australian Journal of Biological Sciences (1952)
Huelin FE