  • Search Results
  • Volatile compounds produced by the reaction of L-cysteine or L-cystine with carbonyl compounds.
Volatile compounds produced by the reaction of L-cysteine or L-cystine with carbonyl compounds.
Agricultural and Biological Chemistry (1973)
Kato S, et al.