  • Search Results
  • Inhibition of pyruvate carboxylase by 1α,25-dihydroxyvitamin D promotes oxidative stress in early breast cancer progression.
Inhibition of pyruvate carboxylase by 1α,25-dihydroxyvitamin D promotes oxidative stress in early breast cancer progression.
Cancer letters (2017-10-13)
Tomasz Wilmanski, Xuanzhu Zhou, Wei Zheng, Aparna Shinde, Shawn S Donkin, Michael Wendt, John R Burgess, Dorothy Teegarden
ABSTRACT
Maintaining reductive-oxidative (redox) balance is an essential feature in breast cancer cell survival, with cellular metabolism playing an integral role in maintaining redox balance through its supply of reduced NADPH. In the present studies, the effect of 1,25-dihydroxyvitamin D (1,25(OH)
Sigma-Aldrich
trans-Dehydroandrosterone, ≥99%