  • Search Results
  • Pulmonary exposure to metal fume particulate matter cause sleep disturbances in shipyard welders.
Pulmonary exposure to metal fume particulate matter cause sleep disturbances in shipyard welders.
Environmental pollution (Barking, Essex : 1987) (2017-10-11)
Hsiao-Chi Chuang, Ting-Yao Su, Kai-Jen Chuang, Ta-Chih Hsiao, Hong-Ling Lin, Yuan-Ting Hsu, Chih-Hong Pan, Kang-Yun Lee, Shu-Chuan Ho, Ching-Huang Lai
ABSTRACT
Sleep disorders may pose a risk to workers in the workplace. We aimed to investigate the associations between metal fume fine particulate matter (PM
Product Description
Supelco
Serotonin, analytical standard