MilliporeSigma
A kaempferol triglycoside from Tephrosia preussii Taub. (Fabaceae).
Natural product research (2017-04-14)
Yves Martial Mba Nguekeu, Maurice Ducret Awouafack, Pierre Tane, Marius Roch Nguedia Lando, Takeshi Kodama, Hiroyuki Morita
ABSTRACT
A phytochemical investigation of the MeOH extract of twigs and leaves of Tephrosia preussi was carried out to give a new kaempferol triglycoside, named tephrokaempferoside (1), together with five known compounds: tephrosin (2), betulinic acid (3), lupeol (4), β-sitosterol (5) and 3-O-β-d-glucopyranoside of β-sitosterol (6). The structure of the new compound was characterised by analyses of NMR (1D and 2D) and MS data, and chemical conversion. Tephrokaempferoside (1) had weak antibacterial activity against Klebsiella pneumoniae with an MIC value of 150 μg/mL.
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG