  • Search Results
  • The daunorubicin interplay with mimetic model membranes of cancer cells: A biophysical interpretation.
The daunorubicin interplay with mimetic model membranes of cancer cells: A biophysical interpretation.
Biochimica et biophysica acta (2017-02-06)
Ana Catarina Alves, Daniela Ribeiro, Miguel Horta, José L F C Lima, Cláudia Nunes, Salette Reis
ABSTRACT
The present work aimed to study the interactions between the anticancer drug daunorubicin and lipid membrane mimetic models of cancer cells composed by their most representative classes of phospholipids, with different degrees of complexity. Regarding these anticancer drug-membrane interactions, several biophysical parameters were assessed using liposomes (LUVs) composed of different molar ratios of DMPC, DOPC, DPPS, DOPE and Chol. In this context, daunorubicin's membrane concentration was determined by calculating its partition coefficient (Kp) between liposomes and water using derivative UV/vis spectrophotometry at 37°C and pH6.3, a typical tumoral microenvironment. Characterization of the zeta potential of such model membranes, in both the absence and presence of the compound, was accomplished through Electrophoretic Light Scattering (ELS). Fluorescence quenching studies, which determine the location of the drug within the bilayer, were carried out using liposomes labelled with DPH and TMA-DPH, fluorescent probes with known membrane position. Temperature dependent steady-state anisotropy assays were also performed to measure the daunorubicin effect on the membranes' microviscosity. The overall results support that daunorubicin permeation depends on the phospholipid membrane composition and causes alterations in the biophysical properties of the bilayers, namely in the membrane fluidity. The interaction of daunorubicin with the studied phospholipids is mainly driven by electrostatic and hydrophobic interactions. These insights demonstrated that not only membranes can affect daunorubicin accumulation in cells but the compound can alter the properties of membranes. The changes produced by daunorubicin on the lipid structure may constitute an additional mechanism of action, which might lead to modifications in the location and, consequently, the activity of membrane signaling proteins.
Sigma-Aldrich
L-α-Phosphatidylethanolamine, dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder