  • Search Results
  • Effects of cultured Cordyceps mycelia polysaccharide A on tumor neurosis factor-α induced hepatocyte injury with mitochondrial abnormality.
Effects of cultured Cordyceps mycelia polysaccharide A on tumor neurosis factor-α induced hepatocyte injury with mitochondrial abnormality.
Carbohydrate polymers (2017-03-08)
Huiling Tang, Weikun Wei, Wang Wang, Zhengqi Zha, Ting Li, Zhijie Zhang, Chen Luo, Hongping Yin, Fengjie Huang, Ying Wang
ABSTRACT
Cordyceps sinensis mycelia polysaccharide A (CPS-A), was isolated from cultured Cordyceps mycelia by 65% alcohol extraction and ion-exchange column chromatography. The molecular weight of CPS-A was 1.2×10
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG