MilliporeSigma
  • MiR-497∼195 cluster regulates angiogenesis during coupling with osteogenesis by maintaining endothelial Notch and HIF-1α activity.
MiR-497∼195 cluster regulates angiogenesis during coupling with osteogenesis by maintaining endothelial Notch and HIF-1α activity.
Nature communications (2017-07-08)
Mi Yang, Chang-Jun Li, Xi Sun, Qi Guo, Ye Xiao, Tian Su, Man-Li Tu, Hui Peng, Qiong Lu, Qing Liu, Hong-Bo He, Tie-Jian Jiang, Min-Xiang Lei, Mei Wan, Xu Cao, Xiang-Hang Luo
ABSTRACT
A specific bone vessel subtype, strongly positive for CD31 and endomucin (CD31
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sigma-Aldrich
Polyvinylpyrrolidone, mol wt (number average molecular weight Mn 360kDa)