  • Search Results
  • Identification of Three Coated Vesicle Components as and
    /3-Tubulin Linked to a Phosphorylated 50,000-dalton
    Polypeptide
Identification of Three Coated Vesicle Components as and<BR/> /3-Tubulin Linked to a Phosphorylated 50,000-dalton<BR/> Polypeptide
The Journal of Cell Biology (1983)
Pfeffer SR