  • Search Results
  • Basal cells as stem cells of the mouse trachea and human airway epithelium.
Basal cells as stem cells of the mouse trachea and human airway epithelium.
Proceedings of the National Academy of Sciences of the USA (2009)
Rock JR