  • Search Results
  • A CO-releasing molecule prevents annexin A2 down-regulation and associated disorders in LPS-administered rat lung.
A CO-releasing molecule prevents annexin A2 down-regulation and associated disorders in LPS-administered rat lung.
Biochemical and biophysical research communications (2017-04-30)
Kana Unuma, Toshihiko Aki, Kanako Noritake, Takeshi Funakoshi, Koichi Uemura
ABSTRACT
To investigate septic lung injuries and the possible relief from injury by carbon monoxide (CO), rats were intraperitoneally (i.p.) administered water or the water-soluble CO-releasing molecule CORM (30 mg/kg body weight), followed by the successive administration of PBS or lipopolysaccharide (LPS, 15 mg/kg body weight, 6 h). The results in four experimental groups (control, LPS, LPS + CORM, CORM, n = 3 or 4 in each groups) were examined. Histological examination revealed the intravascular aggregation of erythrocytes in the lungs of the LPS group, and serological analysis showed a significant increase in D-dimer in the LPS group. Both the aggregation and D-dimer increase were ameliorated in the LPS + CORM group, suggesting that LPS-induced DIC in the lung is ameliorated by CORM. Proteomic as well as immunoblot analyses revealed that the levels of annexin A
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid