  • Search Results
  • Kinetic and mechanistic study on the reactions of ruthenium(ii) chlorophenyl terpyridine complexes with nucleobases, oligonucleotides and DNA.
Kinetic and mechanistic study on the reactions of ruthenium(ii) chlorophenyl terpyridine complexes with nucleobases, oligonucleotides and DNA.
Dalton transactions (Cambridge, England : 2003) (2017-02-01)
Milan M Milutinović, Sofi K C Elmroth, Goran Davidović, Ana Rilak, Olivera R Klisurić, Ioannis Bratsos, Živadin D Bugarčić
ABSTRACT
In this study, we investigated the ability of Ru(ii) polypyridyl complexes to act as DNA binders. The substitution reactions of three Ru(ii) chlorophenyl terpyridine complexes, i.e. [Ru(Cl-Ph-tpy)(en)Cl]Cl (1), [Ru(Cl-Ph-tpy)(dach)Cl]Cl (2) and [Ru(Cl-Ph-tpy)(bpy)Cl]Cl (3) (Cl-Ph-tpy = 4'-(4-chlorophenyl)-2,2':6',2''-terpyridine, en = 1,2-diaminoethane, dach = 1,2-diaminocyclohexane, bpy = 2,2'-bipyridine), with a mononucleotide guanosine-5'-monophosphate (5'-GMP) and oligonucleotides such as fully complementary 15-mer and 22-mer duplexes with a centrally located GG-binding site for DNA, and fully complementary 13-mer duplexes with a centrally located GG-binding site for RNA were studied quantitatively by UV-Vis spectroscopy. Duplex RNA reacts faster with complexes 1-3 than duplex DNA, while shorter duplex DNA (15mer GG) reacts faster compared with 22mer GG duplex DNA. The measured enthalpies and entropies of activation (ΔH
Sigma-Aldrich
Indole-3-butyric acid potassium salt, suitable for plant cell culture, BioReagent
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %
Sigma-Aldrich
Guanosine 5′-monophosphate disodium salt hydrate, from yeast, ≥99%