  • Search Results
  • Genes of ACYL CARRIER PROTEIN Family Show Different Expression Profiles and Overexpression of ACYL CARRIER PROTEIN 5 Modulates Fatty Acid Composition and Enhances Salt Stress Tolerance in
Genes of ACYL CARRIER PROTEIN Family Show Different Expression Profiles and Overexpression of ACYL CARRIER PROTEIN 5 Modulates Fatty Acid Composition and Enhances Salt Stress Tolerance in
Frontiers in plant science (2017-06-24)
Jiexue Huang, Caiwen Xue, Han Wang, Lisai Wang, Wolfgang Schmidt, Renfang Shen, Ping Lan
ABSTRACT
Acyl carrier proteins (ACPs) are a group of small acidic proteins functioning as important cofactors in the
Product Description
Supelco
Supelco 37 Component FAME Mix, certified reference material, TraceCERT®, in dichloromethane (varied conc.), ampule of 1 mL