  • Search Results
  • WINCS Harmoni: Closed-loop dynamic neurochemical control of therapeutic interventions.
WINCS Harmoni: Closed-loop dynamic neurochemical control of therapeutic interventions.
Scientific reports (2017-04-30)
Kendall H Lee, J Luis Lujan, James K Trevathan, Erika K Ross, John J Bartoletta, Hyung Ook Park, Seungleal Brian Paek, Evan N Nicolai, Jannifer H Lee, Hoon-Ki Min, Christopher J Kimble, Charles D Blaha, Kevin E Bennet
ABSTRACT
There has been significant progress in understanding the role of neurotransmitters in normal and pathologic brain function. However, preclinical trials aimed at improving therapeutic interventions do not take advantage of real-time in vivo neurochemical changes in dynamic brain processes such as disease progression and response to pharmacologic, cognitive, behavioral, and neuromodulation therapies. This is due in part to a lack of flexible research tools that allow in vivo measurement of the dynamic changes in brain chemistry. Here, we present a research platform, WINCS Harmoni, which can measure in vivo neurochemical activity simultaneously across multiple anatomical targets to study normal and pathologic brain function. In addition, WINCS Harmoni can provide real-time neurochemical feedback for closed-loop control of neurochemical levels via its synchronized stimulation and neurochemical sensing capabilities. We demonstrate these and other key features of this platform in non-human primate, swine, and rodent models of deep brain stimulation (DBS). Ultimately, systems like the one described here will improve our understanding of the dynamics of brain physiology in the context of neurologic disease and therapeutic interventions, which may lead to the development of precision medicine and personalized therapies for optimal therapeutic efficacy.
Product Description
Supelco
Serotonin, analytical standard