  • Search Results
  • Biosynthetic pathway of aliphatic formates via a Baeyer-Villiger oxidation in mechanism present in astigmatid mites.
Biosynthetic pathway of aliphatic formates via a Baeyer-Villiger oxidation in mechanism present in astigmatid mites.
Proceedings of the National Academy of Sciences of the United States of America (2017-02-23)
Nobuhiro Shimizu, Daisuke Sakata, Eric A Schmelz, Naoki Mori, Yasumasa Kuwahara
ABSTRACT
Astigmatid mites depend on bioactive glandular secretions, pheromones, and defensive agents to mediate intra- and interspecies interactions. Aliphatic formates, such as (
Product Description
Supelco
Bacterial Acid Methyl Ester (BAME) Mix, solution (10 mg/mL total concentration in methyl caproate), analytical standard