  • Search Results
  • Role of steel slags on biomass/carbon dioxide gasification integrated with recovery of high temperature heat.
Role of steel slags on biomass/carbon dioxide gasification integrated with recovery of high temperature heat.
Bioresource technology (2016-10-25)
Yongqi Sun, Qianyi Liu, Hao Wang, Zuotai Zhang, Xidong Wang
ABSTRACT
Disposal of biomass in the agriculture and steel slags in the steel industry provides a significant solution toward sustainability in China. Herein these two sectors were creatively combined as a novel method, i.e., biomass/CO
Sigma-Aldrich
Picropodophyllotoxin, ≥96% (HPLC), powder