Fibroblast growth factor 2 (FGF2) and vascular endothelial growth factor A (VEGFA) synergistically promote steroidogenesis and survival of cultured buffalo granulosa cells.
Animal reproduction science (2017-02-28)
S R Mishra, Jaya Bharati, G Rajesh, V S Chauhan, G Taru Sharma, S Bag, V P Maurya, G Singh, M Sarkar