  • Search Results
  • An historical review of the use of dye precursors in the formulation of commercial oxidation hair dyes.
An historical review of the use of dye precursors in the formulation of commercial oxidation hair dyes.
Dyes and Pigments (1999)
Corbett JF.