  • Search Results
  • A serum-free medium formulation supporting growth of human umbilical cord vein endothelial cells in long-term cultivation.
A serum-free medium formulation supporting growth of human umbilical cord vein endothelial cells in long-term cultivation.
Cytotechnology (2001)
Labitzke R and Friedl P