  • Search Results
  • E-cadherin, MMP-2, and MMP-9 as prognostic markers in penile cancer: analysis of 125 patients.
E-cadherin, MMP-2, and MMP-9 as prognostic markers in penile cancer: analysis of 125 patients.
Urology (2006)
Campos RS et al.