  • Search Results
  • Triton X-100 concentration effects on membrane permeability of a single HeLa cell by scanning electrochemical microscopy (SECM).
Triton X-100 concentration effects on membrane permeability of a single HeLa cell by scanning electrochemical microscopy (SECM).
Proceedings of the National Academy of Sciences of the USA (2010)
Koley D and Bard AJ