  • Search Results
  • Kinetics of Thermal Denaturation and Aggregation of Bovine Serum Albumin.
Kinetics of Thermal Denaturation and Aggregation of Bovine Serum Albumin.
PLoS ONE (2016)
Borzova VA et al.