  • Search Results
  • The newly developed monoclonal antibody SA7D6 exhibits potential for detection of Staphylococcus aureus.
The newly developed monoclonal antibody SA7D6 exhibits potential for detection of Staphylococcus aureus.
Food Science and Biotechnology (2015)
Senevirathne A and Kim K-P