  • Search Results
  • Human umbilical mesenchymal stem cells promote recovery after ischemic stroke.
Human umbilical mesenchymal stem cells promote recovery after ischemic stroke.
Stroke (2011)
Lin YC et al.