  • Search Results
  • TUBA4A gene analysis in sporadic amyotrophic lateral sclerosis: identification of novel mutations.
TUBA4A gene analysis in sporadic amyotrophic lateral sclerosis: identification of novel mutations.
Journal of Neurology (2015)
Pensato V