  • Search Results
  • Fixed single-cell transcriptomic characterization of human radial glial diversity
Fixed single-cell transcriptomic characterization of human radial glial diversity
Nature Methods (2016)
Elliot R Thomsen