  • Search Results
  • Phosphorylated fraction of H2AX as a measurement for DNA damage in cancer cells and potential applications of a novel assay
Phosphorylated fraction of H2AX as a measurement for DNA damage in cancer cells and potential applications of a novel assay
PLoS ONE (2017)
Jiuping Ji