  • Search Results
  • A subset of platinum-containing chemotherapeutic agents kills cells by inducing ribosome biogenesis stress
A subset of platinum-containing chemotherapeutic agents kills cells by inducing ribosome biogenesis stress
Nature Medicine (2017)
Peter M Bruno