  • Search Results
  • A colorimetric method for measurement of the (peroxidase-mediated) oxidation of 3,3'-diaminobenzidine.
A colorimetric method for measurement of the (peroxidase-mediated) oxidation of 3,3'-diaminobenzidine.
The Journal of Histochemistry and Cytochemistry (1973)
Fahimi HD and Herzog V