  • Search Results
  • A new sensitive colorimetric assay for peroxidase using 3,3'-diaminobenzidine as hydrogen donor.
A new sensitive colorimetric assay for peroxidase using 3,3'-diaminobenzidine as hydrogen donor.
Analytical Biochemistry (1973)
Herzog V and Fahimi HD