MilliporeSigma
  • In Vitro Evaluation of Aptamer-Based Reversible Inhibition of Anticoagulant Activated Protein C as a Novel Supportive Hemostatic Approach.
In Vitro Evaluation of Aptamer-Based Reversible Inhibition of Anticoagulant Activated Protein C as a Novel Supportive Hemostatic Approach.
Nucleic acid therapeutics (2016-10-14)
Nasim Shahidi Hamedani, Heiko Rühl, Julia Janina Zimmermann, Tim Heiseler, Johannes Oldenburg, Günter Mayer, Bernd Pötzsch, Jens Müller
ABSTRACT
Activated protein C (APC) is a critical regulator of thrombin formation and thereby protects against thrombosis. On the other hand, overwhelming formation of APC increases the risk of bleeding such as in trauma-induced coagulopathy. Thus, pharmacological inhibition of APC activity may improve blood clottability in certain clinical situations. In this study, we demonstrate that the DNA aptamer HS02-52G binds with fast onset (1.118 ± 0.013 × 10
Sigma-Aldrich
Picropodophyllotoxin, ≥96% (HPLC), powder