  • Search Results
  • Platinum(II) carboxylato complexes containing 7-azaindoles as N-donor carrier ligands showed cytotoxicity against cancer cell lines.
Platinum(II) carboxylato complexes containing 7-azaindoles as N-donor carrier ligands showed cytotoxicity against cancer cell lines.
Journal of inorganic biochemistry (2016-06-29)
Pavel Štarha, Zdeněk Trávníček, Lucia Pazderová, Zdeněk Dvořák
ABSTRACT
The platinum(II) malonato (Mal) and decanoato (Dec) complexes of the general formulas [Pt(Mal)(naza)
Sigma-Aldrich
Guanosine 5′-monophosphate disodium salt hydrate, from yeast, ≥99%