  • Search Results
  • Berry seed press residues and their valuable ingredients with special regard to black currant seed press residues.
Berry seed press residues and their valuable ingredients with special regard to black currant seed press residues.
Food Chemistry (2008)
Helbig, Dorit, et al.